Learn More
TPP™ Tissue Culture Flasks
Surface activated, uniform flat growth surface with crystal clear transparency
Specifications
Material Polystyrene
View More Specs
Description
Description
Surface activated, uniform flat growth surface with crystal clear transparency
  • Surface activated, uniform flat growth surface
  • Crystal clear transparency
  • Geometry of neck reduces risk of media slop over
  • Stable, slip-free stacking of multiple TPP flasks
  • Highly visible bilateral marking area
  • White and accurate volume graduations for optic control of the filling volume
Specifications
Specifications
Polystyrene