missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) chloride, 99%, anhydrous, Thermo Scientific Chemicals

Product Code. 10684702
Click to view available options
Quantity:
10 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-78-8
Molecular Formula Cl4Sn
Molecular Weight (g/mol) 260.52
InChI Key HPGGPRDJHPYFRM-UHFFFAOYSA-J
Synonym tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
PubChem CID 24287
IUPAC Name tetrachlorostannane
SMILES Cl[Sn](Cl)(Cl)Cl

Specifications

Boiling Point 114.0°C
Melting Point -33.0°C
Color Brown-Red
Physical Form Solution
Quantity 1 L
Assay Percent Range 98.5 to 101.5% (Argentometry)
Linear Formula SnCl4
Fieser 01,1111; 03,269; 05,627; 06,553; 07,342; 09,436; 11,522; 12,486; 13,14; 14,304; 15,311; 17,335
Merck Index 15,8901
Solubility Information Solubility in water: reacts. Other solubilities: soluble in carbon disulfide, alcohol, carbon, tetrachloride, benzene, toluene, acetone, kerosene, and gasoline
Formula Weight 260.52
Percent Purity 99%
Packaging Glass bottle
Specific Gravity 2.22
Density 2.2200g/mL
Chemical Name or Material Tin(IV) chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tin(IV) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'