missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog Number p-7050763
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13813-43-9
MFCD00050141
thulium iii iodide, acmc-1boxt
This item is not returnable. View return policy
13813-43-9
MFCD00050141
thulium iii iodide, acmc-1boxt

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 1260°C
Melting Point 1015°C
Physical Form Flakes
Quantity 1 g
Linear Formula TmI3
Formula Weight 549.65
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Thulium(III) iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'