missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7049264
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
13760-79-7
F3Tm
225.93
MFCD00049611
AATUHDXSJTXIHB-UHFFFAOYSA-K
thulium(3+) trifluoride
[F-].[F-].[F-].[Tm+3]
This item is not returnable. View return policy
13760-79-7
F3Tm
225.93
MFCD00049611
AATUHDXSJTXIHB-UHFFFAOYSA-K
thulium(3+) trifluoride
[F-].[F-].[F-].[Tm+3]

Thulium(III) fluoride, anhydrous is used for glass doping and to enhance mid-IR emission in oxyfluoride glass ceramics. It is also used to improve ultraviolet and visible upconversion fluorescence in fluorinated glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 2200°C
Melting Point 1158°C
Physical Form Crystalline
Quantity 1 g
UN Number UN3288
Solubility Information Insoluble in water
Formula Weight 225.93
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 7.9 g/mL
Assay (REO)
Chemical Name or Material Thulium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'