missing translation for 'onlineSavingsMsg'
Learn More

Thiophosgene, 85%, Thermo Scientific Chemicals

Chemical Identifiers

CAS 463-71-8
MDL Number MFCD00004918
InChI Key ZWZVWGITAAIFPS-UHFFFAOYSA-N
Synonym thiophosgene, carbonothioic dichloride, carbon chlorosulfide, thiocarbonyl chloride, thiocarbonic dichloride, dichlorothiocarbonyl, thiofosgen, phosgene, thio, thiokarbonylchlorid, carbonyl chloride, thio
PubChem CID 10040
ChEBI CHEBI:29366
IUPAC Name thiocarbonyl dichloride
SMILES C(=S)(Cl)Cl
View More Specs

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Chemical Identifiers

463-71-8
ZWZVWGITAAIFPS-UHFFFAOYSA-N
10040
thiocarbonyl dichloride
MFCD00004918
thiophosgene, carbonothioic dichloride, carbon chlorosulfide, thiocarbonyl chloride, thiocarbonic dichloride, dichlorothiocarbonyl, thiofosgen, phosgene, thio, thiokarbonylchlorid, carbonyl chloride, thio
CHEBI:29366
C(=S)(Cl)Cl

Specifications

73.0°C
Orange-Red to Red
80% min. (GC)
ClCSCl
03,134
62°C
Solubility in water: slow decomposition in cold, fast in hot water. Other solubilities: soluble in ether, ccl4, decomposes in alcohol
C(=S)(Cl)Cl
114.98
CHEBI:29366
85%
1.508
Thiophosgene
463-71-8
Liquid
CCl2S
MFCD00004918
05,667; 09,466
thiophosgene, carbonothioic dichloride, carbon chlorosulfide, thiocarbonyl chloride, thiocarbonic dichloride, dichlorothiocarbonyl, thiofosgen, phosgene, thio, thiokarbonylchlorid, carbonyl chloride, thio
ZWZVWGITAAIFPS-UHFFFAOYSA-N
thiocarbonyl dichloride
10040
114.98
1.5442
1.5080g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
If skin i

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 207-341-6

Product Identifier
  • Thiophosgene

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only