Learn More
Epredia™ Envi-Ro-Tech™ Freezing Spray
Ozone-friendly freezing spray for use at the microtome and cryostat
Brand: Epredia™ 6769038
Additional Details : CAS Number : 811-97-2 Weight : 2.44000kg Transport : UN number : 3159 Chem class : 2.2
Description
- Facilitates rapid freezing and consistent results
- Attachable capillary nozzle enables spot freezing
- Contains 1,1,1,2 Tetrafluoroethane
Specifications
8 oz, can; 6/Cs | |
Freezing Spray |
Safety and Handling
- Warning
Hazard Category
- Gases under pressure
Hazard Statement
- H280-Contains gas under pressure; may explode if heated.
Precautionary Statement
- P410+P403-Protect from sunlight Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 1,1,1,2- tetraflouroethane