Learn More
Mouse IgG Isotype Control, FITC, Invitrogen™
Mouse Isotype Control
Brand: Invitrogen 31505
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Purity: The precipitin lines against goat anti-mouse whole serum are the same as those against goat anti-mouse IgG, Fc fragment specific when assayed by immunoelectrophoresis at an antigen concentration of 20 mg/mL. No reaction was observed against goat anti-mouse IgA, alpha-chain specific or goat anti-mouse IgM, mu-chain specific. This antibody is recommended for use in blocking assays, immunohistochemical, immunocytochemical and immunofluorescent applications. Fluorophore: Fluorescein-5-isothiocyanate (FITC-isomer 1) Amax = 492 nm; Emax = 520 nm Fluorophore/Protein: 9.3 μg/mg; 3.5 moles FITC per mole IgG Restoration and Storage: Store product at 4°C until opened. Restore with 0.55 mL distilled water (2.0 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. Product may be stored for up to 6 weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of 50% glycerol and store at -20°C Country of Origin: USA
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody's species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.Specifications
Mouse IgG | |
2 mg/mL | |
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
Mouse | |
1 mg | |
Isotype Control (Primary) | |
4° C, store in dark | |
IgG |
Blocking Assay, Control, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry | |
FITC | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; VH7183 | |
purified | |
RUO | |
Not Applicable | |
Lyophilized |