missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SULT1A3, Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ LFP0049
Additional Details : Weight : 0.00010kg
Description
Catecholamine Sulfating Phenol Sulfotranferase Recombinant Protein, LF-P0049, from Invitrogen.
The human Catecholamine-Sulfating Phenol Sulfotransferase (CSPS) is the only sulfotransferase that catalyses the sulfation of catecholamins, in particular the neurotransmitter dopamine, with high activity. CSPS is required for stimulation by Mn2+ of the sulfating activity and expressed in the human intestine, brain, platelet and other tissues. In the brain it may play a role in regulating the levels of dopamine. It also serves as a detoxifying function in the intestine, where it may detoxify potentially lethal dietary monoamines. This protein corresponds to human CSPS (1-295 aa with additional 3 amino acids-GSH at N-term), which has a molecular weight of 36.5 kDa. Purity: >95% by SDS-PAGE.Specifications
P0DMM9 | |
Control | |
6818 | |
7.0 | |
E. coli | |
RUO | |
SULT1A3 | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |
1mg/mL | |
0.02M HEPES with 10% glycerol and no preservative; pH 7.0 | |
Human SULT1A3 | |
250μg | |
-80°C, Avoid Freeze/Thaw Cycles | |
Human SULT1A3 | |
Human | |
Hu | |
Recombinant | |
Recombinant |
in vitro research only