Learn More
Thermo Scientific™ BioBasic™ 8 Javelin™ Guard Columns
Offer convenient, easy-to-use protection for analytical HPLC columns
Brand: Thermo Scientific™ 72205-012106
Code : 16
Additional Details : Weight : 0.10000kg
Description
- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
- 300Å pore size for improved biomolecule analysis
- An excellent starting column for protein and peptide analysis
- Outstanding reproducibility, efficiency and column lifetime
- Excellent for LC/MS separations
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
Specifications
5 μm | |
2.1 mm | |
BioBasic |
10 mm | |
BioBasic 8 |