Learn More
Julabo™ Thermal Bath Fluids
Used for temperature control applications. Julabo™ Thermal Bath Fluids offer safe and reliable operation.
Brand:  Julabo™ 8940120
Code : A0
Additional Details : Weight : 10.00000kg
 View more versions of this product
Product Code. 10518530
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • For closed systems like the Presto or Forte
  • Has wide temperature ranges
  • Low viscosity
  • High stability
  • Good heat conductivity
  • Minimum odor
  • Low corrosion tendency
  • Low toxicity
  • Long shelf life
Specifications
Specifications
Clear
Thermal Bath Fluid
Ideal for temperature control applications and guarantee safe and reliable operation, viscosity, oxi
HL80
>420°C
-85°C to +170°C
Thermal Bath
Drain tap
10 L
3.21mm2/s at 20°C