missing translation for 'onlineSavingsMsg'
Learn More

THEM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen THEM2
Concentration 0.17 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16816524
View Documents Promotion Details
Proteintech
16782-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16806524
View Documents Promotion Details
Proteintech
16782-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

THEM2, also known as ACOT13, belongs to the hotdog-fold superfamily and possesses thioesterase activity, with greater activity observed with longer chain acyl-CoAs such as myristoyl- and palmitoyl-CoA. THEM2 is highly expressed in several tissues such as heart, kidney, liver and brain and has been shown to be co-localized with beta-tubulin on microtubules. THEM2 interacts with StarD, a protein that plays a key role in fatty acid metabolism, and the addition of StarD to THEM2 increased its catalytic activity, suggesting that THEM2 plays a significant role in the metabolism of fatty acids. At least two isoforms of THEM2 are known to exist.
Specifications

Specifications

THEM2
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Human, Rat, Mouse
Q9CQR4, Q9NPJ3
291135, 55856, 66834
THEM2 Fusion Protein Ag10212
Primary
-20°C
ACOT13
0.17 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ACOT13, Acyl CoA thioesterase 13, HT012, PNAS 27, THEM2
Acot13
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications