missing translation for 'onlineSavingsMsg'
Learn More

TGM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15100-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16894114

  • £130.00 / 20µL
Estimated Shipment: 05-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Transglutaminase II catalyzes calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. It is also known as TGC, tTG, type II-, Gh, cytosolic-, liver-, endothelial-, erythrocyte-, cellular-transglutaminase. Different tissues and cell types express varying amounts of tissue transglutaminase with a markedly hugh expression in rheumatoid lesions. It is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.
Specifications

Specifications

TGM2
Polyclonal
Unconjugated
TGM2
G ALPHA h, GNAH, TG(C), TG2, TGase 2, TGase C, TGase H, TGC, TGM2, Tissue transglutaminase, Transglutaminase 2, Transglutaminase C, Transglutaminase H
Rabbit
Antigen Affinity Chromatography
RUO
21817, 56083, 7052
-20°C
Liquid
Immunoprecipitation, Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P21980, P21981
TGM2
TGM2 Fusion Protein Ag7439
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers