missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrachloroethylene, Extra Pure, SLR, Fisher Chemical™
158.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
158.18 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
158.18 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
2.5 L
5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Specifications
Melting Point | -22°C |
Color | Colorless |
Boiling Point | 120°C |
Packaging | Amber glass bottle |
Quantity | 2.5 L |
Formula Weight | 165.83g/mol |
Vapor Pressure | 18mbar at 20°C |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'