missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 4.01 Technical Calibration Buffer Sachets, Hanna Instruments™
Click to view available options
Quantity:
25 x 20 mL
Description
- Premium quality pH 4.01 technical calibration buffer that is NIST traceable and supplied with a Certificate of Analysis
- Accuracy of +/- 0.01 pH @ 25°C
- Temperature chart of the actual pH value at various temperatures printed on each sachet
- HI50004-02 is a premium quality pH 4.01 technical calibration buffer produced in accordance with ISO 3696/BS3978 standards
- Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers
- Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
Specifications
Specifications
Color | Colorless |
pH | 4.01 |
Physical Form | Liquid |
Quantity | 25 x 20 mL |
Solution Type | Buffer |
Formulation | Aqueous Buffer Solution |
Odor | Odorless |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
pH 4.01 Technical Calibration Buffer Sachets, Hanna Instruments™ >
Spot an opportunity for improvement?Share a Content Correction