missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Technical Calibration Buffer pH 10.01, Hanna Instruments™
Click to view available options
Quantity:
20 mL
Specifications
Specifications
Color | Colorless |
pH | 10.01 |
Physical Form | Liquid |
Quantity | 20 mL |
Solution Type | Buffer |
Formulation | Aqueous Buffer Solution |
Odor | Odorless |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Technical Calibration Buffer pH 10.01, Hanna Instruments™ >
Spot an opportunity for improvement?Share a Content Correction