man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Chromatography Systems
Liquid Chromatography Instruments
Individual HPLC Components and Accessories
General HPLC Accessories
Cytiva TC-Gasket
Print
Cytiva TC-Gasket
£222.00 - £397.00
See promotional offers below
Specifications
Connection Type
25 mm
Material
Perfluororubber
Type
TC-Gasket
View More Specs
Products
2
Description
Specifications
Videos
SDS
Documents
Certificates
Products
2
Product Code
Brand
Diameter (Metric) Inner
Price
Quantity & Availability
Product Code
Brand
Diameter (Metric) Inner
Price
Quantity & Availability
11360092
missing translation for 'viewDocuments'
Promotion Details
missing translation for 'viewProductCertificates'
Cytiva
18-1169-24
4 mm
£222.00
Each
In Stock
Log in
to see stock.
Add to basket
11370092
missing translation for 'viewDocuments'
Promotion Details
missing translation for 'viewProductCertificates'
Cytiva
18-1169-25
6.5 mm
£397.00
Each
In Stock
Log in
to see stock.
Add to basket
Description
Description
Specifications
Specifications
Connection Type
25 mm
Material
Perfluororubber
Type
TC-Gasket
Videos
SDS
Documents
Documents
Certificates
Back to Top