missing translation for 'onlineSavingsMsg'
Learn More

Talin-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14168-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16882684

  • £339.00 / 150µL
Estimated Shipment: 05-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene belongs to the splicing factor SR family of genes. U2 auxiliary factor, comprising a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. This gene encodes the small subunit which plays a critical role in both constitutive and enhancer-dependent RNA splicing by directly mediating interactions between the large subunit and proteins bound to the enhancers. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications

Specifications

Talin-1
Polyclonal
Unconjugated
TLN1
ILWEQ, KIAA1027, talin 1, TLN, TLN1
Rabbit
Antigen Affinity Chromatography
RUO
21894, 313494, 7094
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.43 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P26039, Q9Y490
Tln1
Talin-1 Fusion Protein Ag5367
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers