missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sur-8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17561-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
SHOC2 protein participates in protein binding / transferase activity in the fibroblast growth factor receptor signaling pathway and Ras protein signal transduction. It is a widely expressed protein composed almost entirely of leucine-rich repeats (LRR), with a lysine-rich sequence at the amino terminus and cytoplasmically localized. SHOC2 acts as a positive modulator of the RAS-MAPK signaling cascade, which is elicited by EGL-15 and LET-23 and mediated by LET-60. SHOC2 together with protein phosphatase 1c (PP1c) forms a highly specific M-Ras effector complex and is essential for activation of the MAPK pathway by growth factors. Furthermore, in tumor cells with Ras gene mutations, inhibition of SHOC2 expression inhibits MAPK, but not PI3K activity. The SHOC2-PP1c holoenzyme provides an attractive therapeutic target for inhibition of the MAPK pathway in cancer. Recent studies show that aberrantly acquired N-myristoylation of SHOC2 causes human disease Noo-like syndrome with loose anagen hair.Specifications
Sur-8 | |
Polyclonal | |
Unconjugated | |
SHOC2 | |
KIAA0862, Protein soc 2 homolog, Protein sur 8 homolog, SHOC2, SIAA0862, SOC 2, SOC2, SUR 8, SUR8, sur-8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
309548, 56392, 8036 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88520, Q6AYI5, Q9UQ13 | |
Shoc2 | |
Sur-8 Fusion Protein Ag11754 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |