missing translation for 'onlineSavingsMsg'
Learn More

Sur-8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17561-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16867474

  • £339.00 / 150µL
Estimated Shipment: 01-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SHOC2 protein participates in protein binding / transferase activity in the fibroblast growth factor receptor signaling pathway and Ras protein signal transduction. It is a widely expressed protein composed almost entirely of leucine-rich repeats (LRR), with a lysine-rich sequence at the amino terminus and cytoplasmically localized. SHOC2 acts as a positive modulator of the RAS-MAPK signaling cascade, which is elicited by EGL-15 and LET-23 and mediated by LET-60. SHOC2 together with protein phosphatase 1c (PP1c) forms a highly specific M-Ras effector complex and is essential for activation of the MAPK pathway by growth factors. Furthermore, in tumor cells with Ras gene mutations, inhibition of SHOC2 expression inhibits MAPK, but not PI3K activity. The SHOC2-PP1c holoenzyme provides an attractive therapeutic target for inhibition of the MAPK pathway in cancer. Recent studies show that aberrantly acquired N-myristoylation of SHOC2 causes human disease Noo-like syndrome with loose anagen hair.
Specifications

Specifications

Sur-8
Polyclonal
Unconjugated
SHOC2
KIAA0862, Protein soc 2 homolog, Protein sur 8 homolog, SHOC2, SIAA0862, SOC 2, SOC2, SUR 8, SUR8, sur-8
Rabbit
Antigen Affinity Chromatography
RUO
309548, 56392, 8036
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88520, Q6AYI5, Q9UQ13
Shoc2
Sur-8 Fusion Protein Ag11754
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers