Learn More
Glas-Col Support for Flexible Fabric Heating Mantle
This Flexible Fabric Heating Mantle Support safely support flasks and mantles.
Brand:  Glas-Col 36225-84
Product Code. 11786098
  • £156.00 / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Free up valuable bench space while heating solutions.
  • Use these extension supports for attaching hemispherical and spherical mantle to lattice.
  • Each extension is made of a steel ring and heat resistant strap basket.
Specifications
Specifications
Steel
Support
1 Ea.
1000 mL Flasks
For Glass-Col Flexible Fabric Heating Mantle