Learn More
Sulfurous acid, ACS, 6.0% SO{2} min, Thermo Scientific Chemicals
£64.92 - £244.22
Chemical Identifiers
CAS | 7782-99-2 |
---|---|
Molecular Formula | H2O3S |
Molecular Weight (g/mol) | 82.073 |
MDL Number | MFCD00011335 |
InChI Key | LSNNMFCWUKXFEE-UHFFFAOYSA-N |
Synonym | sulphurous acid, sulfur dioxide solution, schweflige saure german, h2so3, unii-j1p7893f4j, schweflige saure, acide sulfureux, acido sulfuroso, schweflige saeure, trioxosulfuric acid |
PubChem CID | 1100 |
ChEBI | CHEBI:48854 |
IUPAC Name | sulfurous acid |
SMILES | OS(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11434990
|
Thermo Scientific Alfa Aesar
033275.AP |
500 mL |
£64.92
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11444990
|
Thermo Scientific Alfa Aesar
033275.AY |
4 x 500 mL |
£244.22
2000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sulfurous acid is used as a reducing agent, bleaching agent and disinfectant. It is utilized for bleaching of moistened wool, straw for hats, and other materials injured by chlorine. It finds application as a disinfectant as well as used externally for its parasiticidal effect in various skin diseases. Its derivative viz. sulfites are included in antiseptics, antifermentatives, and antizymotics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7782-99-2 | |
82.073 | |
LSNNMFCWUKXFEE-UHFFFAOYSA-N | |
1100 | |
sulfurous acid |
H2O3S | |
MFCD00011335 | |
sulphurous acid, sulfur dioxide solution, schweflige saure german, h2so3, unii-j1p7893f4j, schweflige saure, acide sulfureux, acido sulfuroso, schweflige saeure, trioxosulfuric acid | |
CHEBI:48854 | |
OS(=O)O |
Specifications
Colourless liquid | |
7782-99-2 | |
Pungent | |
H2O3S | |
MFCD00011335 | |
Miscible with water. | |
OS(=O)O | |
82.073 | |
CHEBI:48854 | |
ACS |
Colorless | |
1.03 g/cm3 at 20°C | |
Air Sensitive | |
Liquid | |
sulphurous acid, sulfur dioxide solution, schweflige saure german, h2so3, unii-j1p7893f4j, schweflige saure, acide sulfureux, acido sulfuroso, schweflige saeure, trioxosulfuric acid | |
LSNNMFCWUKXFEE-UHFFFAOYSA-N | |
sulfurous acid | |
1100 | |
82.08 | |
Sulfurous acid, 6.0% SO2 min. |
Safety and Handling
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: II
missing translation for 'einecsNumber' : 231-973-1
missing translation for 'rtecsNumber' : WT2775000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Sulphurous acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Sulfurous acid
RUO – Research Use Only