missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid, 0.2N Standardized Solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
4 L
20 L
Description
Sulfuric acid, 0.2N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Quantity | 4 L |
Physical Form | Liquid |
Solubility Information | Miscible with water. |
Formula Weight | 98.08 |
Chemical Name or Material | Sulfuric acid, 0.2N Standardized Solution |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'