Learn More
Fisherbrand™ SUB-GEL Teaching Horizontal Gel Tank Package
SUB-GEL Teaching gel tanks are ideal for demonstration of horizontal gel electrophoresis, helps teachers to demonstrate horizontal gel electrophoresis to many students using just single gel tank and power supply.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.80000kg
Description
- High quality injection moulded construction
- Leak free ‘Plug-and-Go’ casting dams
- UV transparent gel trays
- Height adjustable combs
- Low gel and buffer volumes
- Cassette-style electrodes - difficult to break and easy to change
- Each tank is available with or without power supply
Specifications
Twelve x 8 sample, 1mm MC (2 combs per gel) | |
Applications: Education - ideal for demonstrating horizontal gel electrophoresis to student classes, high throughput electrophoresis of mini gels | |
7 x 7 cm (2.7 x 2.7 in.) | |
96 samples with standard comb option, 196 samples using optional 16 sample combs | |
39.5 x 23 x 9 cm (15.5 x 9 x 3.5 in.) |
Maximum Gel throughput: Six 70mm x 70mm mini Gels | |
6X Mini Classroom Workstation | |
Four SUB-GEL 6X Mini units (Cat. No 15321357) and power supply (Cat. No 12613546) | |
1200 mL |