missing translation for 'onlineSavingsMsg'
Learn More

Strontium fluoride, Optical Grade, Thermo Scientific™

£35.86 - £405.00

Chemical Identifiers

CAS 7783-48-4
Molecular Formula F2Sr
Molecular Weight (g/mol) 125.62
MDL Number MFCD00011251
InChI Key FVRNDBHWWSPNOM-UHFFFAOYSA-L
Synonym strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g
PubChem CID 82210
SMILES [F-].[F-].[Sr++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11371628
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
011020.04
2 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11381628
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
011020.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11361628
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
011020.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7783-48-4
125.62
FVRNDBHWWSPNOM-UHFFFAOYSA-L
82210
F2Sr
MFCD00011251
strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g
[F-].[F-].[Sr++]

Specifications

1473°C
Broken Crystals
MFCD00011251
strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g
[F-].[F-].[Sr++]
125.62
125.62
Odorless
4.240 g/mL
7783-48-4
F2Sr
14,8842
FVRNDBHWWSPNOM-UHFFFAOYSA-L
strontium(2+) difluoride
82210
Optical
1.442
Strontium fluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 232-000-3

missing translation for 'rtecsNumber' : WK8925000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates