missing translation for 'onlineSavingsMsg'
Learn More

Strontium chloride, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407962
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
10476-85-4
SrCl2
MFCD00011249
This item is not returnable. View return policy
10476-85-4
SrCl2
MFCD00011249

Strontium chloride  is used as a source of redness in fireworks. Strontium chloride is often used as a red coloring agent in pyrotechnics. It is employed in small quantities in glass-making and metallurgy. The radioactive isotope strontium-89, used for the treatment of bone cancer, is usually administered in the form of strontium chloride. Sea water aquaria require small amounts of strontium chloride, which is consumed in the production of the exoskeletons of certain plankton. SrCl2 is useful in reducing tooth sensitivity by forming a barrier over microscopic tubules in the dentin containing nerve endings that have become exposed by gum recession. strontium chloride exposure induces parthenogenetic activation of oocytes[6] which is used in developmental biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1250°C
Melting Point 873°C
Physical Form Beads
Quantity 5 g
Assay Percent Range 99.995% (metals basis)
Solubility Information It is soluble in water,slightly soluble in ethanol,acetone. Insoluble in ammonia.
Formula Weight 158.53
Refractive Index 1.65
Sensitivity Hygroscopic
Density 3.052
Chemical Name or Material Strontium chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'