Learn More
Astell Scientific™ Strip Chart Recorder, Two Pen Temperature
Record autoclave cycling temperatures with this strip chart recorder. Astell Scientific™ Strip Chart Recorder, Two Pen Temperature records both chamber and load temperature.
Brand: Astell Scientific™ AAR023
Additional Details : Weight : 0.01000kg
Description
- Two-pen system.
- Records both chamber and load temperature.
Specifications
Swiftlock top loading autoclaves | |
Strip Chart Recorder | |
Two-pen, chamber and load temperature |