Learn More
Astell Scientific™ Strip Chart Recorder, Three Pen, Temperature/Pressure
Record autoclave cycling temperatures and pressure with this strip chart recorder. Astell Scientific™ Strip Chart Recorder, Three Pen, Temperature/Pressure records chamber, load temperature, and chamber pressure.
Brand: Astell Scientific™ AAR091
Additional Details : Weight : 0.01000kg
Description
- Three-pen system.
- Records chamber temperature, load temperature and chamber pressure.
Specifications
Swiftlock top loading autoclaves | |
Strip Chart Recorder | |
Three-pen, chamber temperature, load temperature and chamber pressure |