Learn More
IBA Lifesciences Streptamer™ CD3/CD28 Premix for T Cell Expansion
Streptamer CD3/CD28 Premix for T Cell Expansion is for reversible CD3 and CD28 receptor specific cell activation/expansion using the Streptamer technology.
Brand:  IBA Lifesciences 6-8901-000
Product Code. 15633988
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • For 1.6 x 107 cells in total
  • The vial contains CD3 Fab-Strep, CD28 Fab-Strep and multimeric Strep-Tactin™ for cell expansion, premixed and ready to use, in a total volume of 480 μL
  • With this ready-to-use reagent you will benefit from a quick protocol
  • The multimers can be removed from the cells by the addition of D-Biotin
  • Suspended in sterile PBS
  • Product is not classified as hazardous according to (EC) No 1272/2008 [CLP]
  • Solution is sterile filtered and does not contain any preservatives
  • Please handle under sterile conditions
Specifications
Specifications
480 μL
2°C to 8°C
For stimulation of 1.6 x 107 cells