Learn More
Sampling Systems™ SteriWare™ Single Use Production and Sampling Scoops
These SteriWare™ Single Use Production and Sampling powder Scoops are ideal for sampling, production and laboratory work. The complete range is made to the highest standards.
£71.30 - £174.00
Specifications
Material | Polystyrene |
---|---|
Disposable | Yes |
Packaging Type | Individually Bagged |
Product Code | Brand | Capacity (Metric) | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Sterility | Price | Quantity & Availability | ||||
17524626
|
Sampling Systems™
8286A-25 |
25 mL | Non-sterile |
£71.30
Pack of 100 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17554626
|
Sampling Systems™
8286A-25S |
25 mL | Sterile |
£95.55
Pack of 100 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17584626
|
Sampling Systems™
8286A-50S |
50 mL | Non-sterile |
£106.30
Pack of 100 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17564616
|
Sampling Systems™
8286A-100 |
100 mL | Non-sterile |
£93.85
Pack of 100 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17594616
|
Sampling Systems™
8286A-100S |
100 mL | Sterile |
£124.00
Pack of 100 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17504626
|
Sampling Systems™
8286A-150 |
150 mL | Non-sterile |
£102.00
Pack of 100 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17514626
|
Sampling Systems™
8286A-150S |
150 mL | Sterile |
£138.00
Pack of 100 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17534626
|
Sampling Systems™
8286A-250 |
250 mL | Non-sterile |
£112.00
Pack of 100 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17544626
|
Sampling Systems™
8286A-250S |
250 mL | Sterile |
£164.00
Pack of 100 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17564626
|
Sampling Systems™
8286A-500 |
500 mL | Non-sterile |
£107.00
Pack of 40 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17574626
|
Sampling Systems™
8286A-500S |
500 mL | Sterile |
£161.00
Pack of 40 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17574616
|
Sampling Systems™
8286A-1000 |
1000 mL | Non-sterile |
£136.00
Pack of 30 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17584616
|
Sampling Systems™
8286A-1000S |
1000 mL | Sterile |
£174.00
Pack of 30 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
Description
- SteriWare™ flat bottomed powder scoops are single use, free-standing scoops designed for use in GMP manufacturing areas.
- They are perfect for sampling and dispensing powders and granules.
- These powder scoops are manufactured in a cleanroom to the highest standards.
- Everything from the ergonomic design to the class leading documentation means these are the ideal scoop for use in pharmaceutical and food factories.
- The robust design and ergonomic handle ensures that they are easy and comfortable to use.
- All powder scoops are individually packed and are available in both sterile and non-sterile versions.
- Simply choose your scoop size, unpack and use.
- SteriWare™ Scoops can be used as volumetric scoops by levelling off the excess powder to give you the stated set volume.
- They are available in a wide ranges of sizes from 25 mL (0.9 oz.) to 1000 mL (36 oz.).
- Use straight from the bag.
- The robust, ergonomic handles has been designed so that they can be used repeatedly.
- If measuring powder after scooping remove the excess powder to give an accurate volume.
- Made from food grade PS.
- Conforms to the latest FDA and EU requirements.
- BSE/TSE free.
Specifications
Polystyrene | |
Yes | |
Individually Bagged |