Learn More
Invitrogen™ STAT5 alpha/beta Human InstantOne™ ELISA Kit
InstantOne ELISA
Brand: Invitrogen™ 85-86111-11
Additional Details : Weight : 0.47000kg
Description
The InstantOne™ ELISA is specifically engineered for accurate measurement of total human STAT5A/B in cell lysates. Detection of mouse and rat STAT5A/B is expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
Carries out a dual function: signal transduction and activation of transcription. Mediates cellular responses to the cytokine KITLG/SCF and other growth factors. Mediates cellular responses to ERBB4. May mediate cellular responses to activated FGFR1, FGFR2, FGFR3 and FGFR4. Binds to the GAS element and activates PRL-induced transcription. Regulates the expression of milk proteins during lactation.Specifications
P42229,P51692 | |
ELISA Kit | |
Human | |
6776, 6777 | |
Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal | |
96 Tests | |
Cell Lysate,20 μL | |
Human | |
1 hr. 10 min. |
Biotin | |
Cell Lysates | |
Colorimetric Microplate Reader | |
STAT5,MGF | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |