missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences StarGate pPSG-IBA Acceptor Vectors for E. coli Expression
Achieves high-level expression in E. coli by the T7 promoter by providing specified genetic surroundings for optimal expression and purification of non-toxic proteins.
Brand: IBA Lifesciences 5-4364-001
This item is not returnable.
View return policy
Description
pPSG-IGA E. coli expression vectors:
pPSG-IBA vectors use the T7 promoter and T7 RNA polymerase for high-level transcription of the gene of interest.
- High-level expression in E. coli by bacteriophage T7 promoter
- High-level transcription by T7 RNA polymerase in BL21 strains
- High-level expression of non-toxic proteins
- Induction by IPTG
- Suitable for in vitro transcription/translation
StarGate System
IBA developed StarGate to provide a simple, fast and highly efficient cloning process for genes of interest (GOI) for use in functional expression, purification, detection, immobilization or separation of recombinant proteins:
- Minimal extra modification of the gene due to short combinatorial sites
- Inherent high-level cloning efficiency due to a directed reaction (no equilibrium)
- GOIs are cloned to obtain a Donor Vector; 2 or more genes can be combined using Fusion Cloning
- Genes are then transferred into Acceptor Vectors that provide optimal genetic surroundings
- The final expression vector, the Destination Vector, is then placed into the respective host
Specifications
Liquid | |
pPSG-IBA 164 | |
8 | |
Acceptor Vector for E. coli expression | |
2°C to 8°C for frequent usage, -20°C for long-term storage | |
250 ng/μL | |
Total length: 3469 | |
Cloning genes of interest for use in functional expression, purification, detection, immobilization or separation of recombinant proteins |
StarGate™ System, E. Coli BL21(DE3) | |
E. coli host with T7 RNA polymerase gene (e.g. BL21(DE3)) | |
T7 | |
20 μL TE buffer: 10 mm Tris-HCl, 1 mm EDTA / 5 μg | |
Twin-Strep-tag for N-terminal and FLAG-tag for C-terminal | |
5 μg | |
Clear |