Learn More
Thermo Scientific™ Standard Plate Count Agar (APHA) (Dehydrated)
Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this medium.
Brand: Thermo Scientific™ CM0463T
This item is not returnable.
View return policy
Description
Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this medium.
- Meets formulation of APHA, AOAC and PHLS
- Quality control of Standard Plate Count Agar (APHA) includes testing in accordance with ISO 11133:2014
Specifications
APHA, AOAC, PHLS, ISO 11133:2014 | |
Agar | |
5 kg |
Powder | |
Standard Plate Count for Microorganism |