Learn More
Nickel Electro™ Stainless Steel Gable Lid
Maintain stability in bath temperature and reduce evaporation with a stainless steel lid. Nickel Electro™ Clifton™ Stainless Steel Gable Lid minimises heat loss and reduces evaporation.
Brand:  Nickel Electro™ SL4-14
Code : 03
Additional Details : Weight : 1.14300kg
 View more versions of this product
Product Code. 11469519
  • £268.00 / Each
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
  • Stainless steel
  • Insulated handle
  • Minimises heat loss
  • Reduces evaporation
  • Assists in maintaining bath temperature
Specifications
Specifications
Stainless steel gable lid
14 L Stirred Bath
Stainless Steel
3 Years