Learn More
OHAUS™ Stainless Steel Lab Frame
OHAUS™ Stainless Steel Lab Frame provides secure support for holding glassware at elevated positions.
£845.00 - £3270.00
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15772369
|
OHAUS™
30392323 |
610 x 457 x 610 mm (H x L x W) |
£845.00
Each |
In Stock
Log in to see stock. |
|||||
15782369
|
OHAUS™
30392324 |
610 x 457 x 1219 mm (H x L x W) |
£1040.00
Each |
In Stock
Log in to see stock. |
|||||
15792369
|
OHAUS™
30392325 |
1219 x 457 x 1219 mm (H x L x W) |
£1455.00
Each |
In Stock
Log in to see stock. |
|||||
15702379
|
OHAUS™
30392326 |
1219 x 305 x 1829 mm (H x L x W) |
£1590.00
Each |
In Stock
Log in to see stock. |
|||||
15722379
|
OHAUS™
30392328 |
1905 x 551 x 1829 mm (H x L x W) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Dependable connectors secure rods and clamps to meet your requirements and five unique holders in a variety of sizes and materials accommodate different rods.
Specifications
Lab Frame Kit | |
Stainless Steel |