Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class F1
Calibrate masses of class II and III weighing instruments. Fisherbrand™ Class F1 Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy.
Brand:  Fisherbrand™ FB67822
 View more versions of this product
Product Code. 15515109
  • £722.00 / 1 unit
Estimated Shipment: 11-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class F1 stainless steel calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
F1
1 to 100 g
1 g to 100 g
Cylindrical
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class F1 > 1g to 100g