Learn More
SI Analytics™ Stainless Steel Brackets
Protect your viscometer from breakage. SI Analytics™ Stainless Steel Brackets are designed to ensure that the viscometers are held in a vertical position.
POA
Specifications
Product Type | Bracket |
---|---|
Material | Stainless Steel |
For Use With (Application) | For manual and automatic measurements |
Description
- The maximum deviation is < 1°
- In application in conjunction with SCHOTT Instruments and other commercially available see-through thermostats the viscometers can only be used with the appropriate stand or bracket
- Provide protection for viscometers
- Allow viscometers to be held vertically
Specifications
Bracket | |
Stainless Steel | |
For manual and automatic measurements |