missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRP19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | SRP19 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16835494
|
Proteintech
16033-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16825494
|
Proteintech
16033-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is a rabbit polyclonal antibody raised against full length SRP19 of human origin.
SRP19 belongs to the SRP19 family. It is signal-recognition-particle assembly and binds directly to 7S RNA and mediates binding of the 54 kDa subunit of the SRP.Specifications
SRP19 | |
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P09132, Q9D7A6 | |
291685, 66384, 6728 | |
SRP19 Fusion Protein Ag8903 | |
Primary | |
-20°C | |
SRP19 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
SRP19 | |
Srp19 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |