missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Split/Splitless Liners for Agilent™ 6890 GCs: Low Pressure Drop

Includes wool packing

Brand:  Restek™ 20995

Code : Z3

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 13194332

  • £192.39 / Pack of 5
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

  • I.D. 4mm

Specifications

Specifications

IP deactivation
IP Low Pressure Drop Inlet Liner
7.85 cm
0.63 cm
Borosilicate Glass
Deactivated Wool
5/Pk.
0.4 cm
Agilent GCs Equipped with Split/Splitless Inlets
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers