Learn More
3M™ Soy Protein ELISA Kit, E96SOY
Intended for the detection of soy proteins in clean-in-place water (CIP) final rinse water, environmental swab samples, food ingredients and processed food products.
Brand:  3M™ 7100151351
Product Code. 17233814
  • £864.00 / 1 set
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed to detect both processed and unprocessed target allergen proteins.
  • Test can accommodate environmental swab, CIP rinse water and food samples.
  • A universal extraction protocol streamlines testing for multiple analytes, saving you reagents and valuable time.
Specifications
Specifications
Test
96 Wells
50 Minute