Learn More
Sonoco™ ThermoSafe InsulatedShipper-PUR, Packed in Pallet
High performance polyurethane containers meet the needs of the most demanding temperature sensitive payloads
Brand:  Sonoco™ E65
Additional Details : Weight : 3.50000kg
 View more versions of this product
Product Code. 16308056
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Polyurethane insulation offers greater insulating power than equivalent thicknesses of EPS containers. Thick walls and 200 lb. outer corrugate offer protective shock absorption and cushioning during shipping and handling.
  • Protects highly temperature sensitive shipments from moderate and extreme ambient temperature exposure
  • Provides extensive protection for long range journeys
  • Durable one-piece closed-cell polyurethane foam insulated shippers are CFC and HCFC-free—comply with international clean air regulations
  • Proprietary process laminates highly efficient insulation core to inner and outer corrugate walls
  • Withstands crush compression in excess of 5000 lb.
  • Flexible polyurethane foam plug seals the top, cushions and insulates contents, vents dry ice
  • R-value is +1.5 times EPS R-value, per inch of thickness
Specifications
Specifications
43.8 cm
47.6 cm
12.75 in.
Protects highly temperature sensitive shipments from moderate and extreme ambient temperature exposure
35.5 cm
20.3 cm
25.4 cm
17.25 in.
18.75 in.
18.75 x 12.75 x 17.25 in. (47.6 x 32.3 x 43.8 cm)
14 in.
32.3 cm
10 in.