Learn More
Sodium-silica gel, 35-40% alkali metal in silica gel, Stage I, Thermo Scientific™
Reductively Produces Radical Anions and Dianions, Dechlorinates Alkyl and Aryl Halides and Desulfurize Compounds
Brand: Thermo Scientific Alfa Aesar 044893.06
Code : NEW
Additional Details : Weight : 0.00500kg Transport : UN number : 2813 Chem class : 4.3 Pack group : II
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
Sodium-silica gel | |
5 g | |
UN2813 | |
35-40% alkali metal in silica gel | |
Moisture sensitive | |
0.7 |
Powder | |
MFCD01941718 | |
Reacts with water. | |
Glass bottle | |
0.7 |