missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Dibasic, Anhydrous, Multi-Compendial, U.S.P., J.T. Baker™
179.77 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
4 x 500 g
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
CAS | 7558-79-4 |
Quantity | 4 x 500 g |
Molecular Formula | HNa2O4P |
MDL Number | MFCD00003496 |
Synonym | disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt |
InChI Key | BNIILDVGGAEEIG-UHFFFAOYSA-L |
SMILES | [Na+].[Na+].OP([O-])([O-])=O |
IUPAC Name | disodium;hydrogen phosphate |
Molecular Weight (g/mol) | 141.96 |
PubChem CID | 24203 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'