missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Dibasic, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
246.05 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Description
- Suitable for Buffer Solutions
Specifications
Quantity | 500 g |
IUPAC Name | disodium;hydrogen phosphate |
Formula Weight | 141.96 |
Grade | Meets ACS reagent chemical requirements |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.07kg |
Chemical Name or Material | Sodium Phosphate Dibasic, Anhydrous |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'