missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitrite, Certified AR for Analysis, Fisher Chemical™
£58.15 - £427.00
Chemical Identifiers
CAS | 7632-00-0 |
---|---|
Molecular Formula | NNaO2 |
Molecular Weight (g/mol) | 69.00 |
MDL Number | MFCD00011118 |
InChI Key | LPXPTNMVRIOKMN-UHFFFAOYSA-M |
Synonym | sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts |
PubChem CID | 23668193 |
ChEBI | CHEBI:78870 |
IUPAC Name | sodium;nitrite |
SMILES | [Na+].[O-]N=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10498990
|
Fisher Chemical
S/5640/53 |
500 g |
£58.15
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10274110
|
Fisher Chemical
S/5640/62 |
2.5 kg |
£427.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium Nitrite, Certified AR for Analysis
Chemical Identifiers
7632-00-0 | |
69.00 | |
LPXPTNMVRIOKMN-UHFFFAOYSA-M | |
23668193 | |
sodium;nitrite |
NNaO2 | |
MFCD00011118 | |
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts | |
CHEBI:78870 | |
[Na+].[O-]N=O |
Specifications
320°C | |
7632-00-0 | |
8 to 9 | |
NNaO2 | |
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts | |
[Na+].[O-]N=O | |
69.00 | |
CHEBI:78870 | |
Plastic powder jar |
271°C | |
Yellow | |
Solid | |
MFCD00011118 | |
LPXPTNMVRIOKMN-UHFFFAOYSA-M | |
sodium;nitrite | |
23668193 | |
69g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium nitrite
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 3
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
Hazard Statement
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.