missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitrate, puriss. p.a. ACS Reagent, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO,≥99.5%
£47.40 - £217.60
Chemical Identifiers
CAS | 7631-99-4 |
---|---|
Molecular Formula | NNaO3 |
Molecular Weight (g/mol) | 84.99 |
MDL Number | MFCD00011119 |
InChI Key | VWDWKYIASSYTQR-UHFFFAOYSA-N |
Synonym | sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1 |
PubChem CID | 24268 |
ChEBI | CHEBI:63005 |
IUPAC Name | sodium;nitrate |
SMILES | [Na+].[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15683420
|
Honeywell-Fluka
31440-100G |
100 g |
£64.14
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15603430
|
Honeywell-Fluka
31440-500G |
500 g |
£47.40
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15693420
|
Honeywell-Fluka
31440-1KG |
1 kg |
£75.26
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15623430
|
Honeywell-Fluka
31440-6X500G |
6 x 500 g |
£217.60
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7631-99-4 | |
84.99 | |
VWDWKYIASSYTQR-UHFFFAOYSA-N | |
24268 | |
sodium;nitrate |
NNaO3 | |
MFCD00011119 | |
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1 | |
CHEBI:63005 | |
[Na+].[O-][N+]([O-])=O |
Specifications
7631-99-4 | |
NaNO3 | |
1498 | |
VWDWKYIASSYTQR-UHFFFAOYSA-N | |
sodium;nitrate | |
24268 | |
84.99g/mol | |
ACS/puriss. p.a. |
NNaO3 | |
MFCD00011119 | |
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1 | |
[Na+].[O-][N+]([O-])=O | |
84.99 | |
CHEBI:63005 | |
≥99.5% | |
Sodium Nitrate, Reag. ISO |
Safety and Handling
P220-P305 + P351 + P338
H272-H319
missing translation for 'einecsNumber' : 231-554-3
missing translation for 'rtecsNumber' : WC5600000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium nitrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Sodium nitrate