missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium iodide, Honeywell Fluka™
ACS Reagent,≥99.5%
£67.97 - £3382.60
Chemical Identifiers
CAS | 7681-82-5 |
---|---|
Molecular Formula | INa |
Molecular Weight (g/mol) | 149.894 |
MDL Number | MFCD00003532 |
InChI Key | FVAUCKIRQBBSSJ-UHFFFAOYSA-M |
Synonym | sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech |
PubChem CID | 5238 |
ChEBI | CHEBI:33167 |
IUPAC Name | sodium;iodide |
SMILES | [Na+].[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15695300
|
Honeywell-Fluka
383112-100G |
100 g |
£67.97
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15625310
|
Honeywell-Fluka
383112-500G |
500 g |
£247.32
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15615310
|
Honeywell-Fluka
383112-2.5KG |
2.5 kg |
£843.41
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15605310
|
Honeywell-Fluka
383112-12KG |
12 kg |
£3382.60
12kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7681-82-5 | |
149.894 | |
FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
5238 | |
sodium;iodide |
INa | |
MFCD00003532 | |
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
CHEBI:33167 | |
[Na+].[I-] |
Specifications
7681-82-5 | |
NaI | |
UN3077 | |
FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
sodium;iodide | |
5238 | |
149.89g/mol | |
Vapor Density: >1 (vs air) |
INa | |
MFCD00003532 | |
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
[Na+].[I-] | |
149.894 | |
CHEBI:33167 | |
≥99.5% | |
Sodium iodide |
Safety and Handling
P273-P305 + P351 + P338
missing translation for 'rtecsNumber' : WB6475000