missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, Volumetric, 0.5 M NaOH (0.5N), Honeywell Fluka™

Catalog Number p-7111813
Click to view available options
Quantity:
500 mL
2 L
Packaging:
Poly Bottle
1310-73-2, 7732-18-5
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
1310-73-2, 7732-18-5
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

Volumetric, 0.5 M NaOH (0.5N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY
Boiling Point >100°C
CAS Min % ≥90.0000%, ≥1.0000%
CAS Max % ≤100.0000%, <2.0000%
Linear Formula NaOH
Concentration 0.5 M NaOH (0.5N)
UN Number UN1824
Density 1.02 g/mL at 25 °C
Quantity 500 mL
Formula Weight 40.00
Grade Volumetric
Packaging Poly Bottle
Chemical Name or Material Sodium hydroxide solution, Volumetric, 0.5 M NaOH (0.5N)
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : hydroxyde de sodium
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'