missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution, 0.1M, Honeywell Fluka™

Product Code. 15603580
Click to view available options
Quantity:
500 mL
1 L
2 L
2.5 L
6 x 500 mL
6 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Volumetric, 0.1 M NaOH (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
Concentration 0.1M
UN Number NONH for all modes of transport
Density 1g/mL at 25°C
Quantity 6 x 1 L
Formula Weight 40g/mol
Vapor Density >1 (vs air)
Vapor Pressure 3mmHg (37°C)
Chemical Name or Material Sodium Hydroxide Solution
compliance-icons
Product Identifier
  • Sodium hydroxide solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide; caustic soda
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'