missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Monohydrate, Honeywell Fluka™
TraceSELECT™, for trace analysis,≥99.9995% (metals basis)
£69.74 - £219.50
Chemical Identifiers
CAS | 12200-64-5 |
---|---|
Molecular Formula | H3NaO2 |
Molecular Weight (g/mol) | 58.012 |
MDL Number | MFCD00149229 |
InChI Key | JBJWASZNUJCEKT-UHFFFAOYSA-M |
Synonym | sodium hydroxide monohydrate, na.ho.h2o, naoh water, water naoh, sodium hydroxide h2o, sodium hydroxide water, water sodium hydroxide, h2o naoh, naoh h2o, sodium,hydroxide,hydrate |
PubChem CID | 23719242 |
IUPAC Name | sodium;hydroxide;hydrate |
SMILES | O.[OH-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15600560
|
Honeywell-Fluka
01968-25G |
25 g |
£69.74
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15690550
|
Honeywell-Fluka
01968-100G |
100 g |
£219.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Chemical Identifiers
12200-64-5 | |
58.012 | |
JBJWASZNUJCEKT-UHFFFAOYSA-M | |
23719242 | |
O.[OH-].[Na+] |
H3NaO2 | |
MFCD00149229 | |
sodium hydroxide monohydrate, na.ho.h2o, naoh water, water naoh, sodium hydroxide h2o, sodium hydroxide water, water sodium hydroxide, h2o naoh, naoh h2o, sodium,hydroxide,hydrate | |
sodium;hydroxide;hydrate |
Specifications
12200-64-5 | |
NaOH · H2O | |
1823 | |
JBJWASZNUJCEKT-UHFFFAOYSA-M | |
sodium;hydroxide;hydrate | |
23719242 | |
≥99.99% (metals basis) | |
Sodium Hydroxide Monohydrate |
H3NaO2 | |
MFCD00149229 | |
sodium hydroxide monohydrate, na.ho.h2o, naoh water, water naoh, sodium hydroxide h2o, sodium hydroxide water, water sodium hydroxide, h2o naoh, naoh h2o, sodium,hydroxide,hydrate | |
O.[OH-].[Na+] | |
58.012 | |
58.01g/mol | |
TraceSELECT™ |
Safety and Handling
P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H314
missing translation for 'einecsNumber' : 215-185-5
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium hydroxide monohydrate
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Sodium hydroxide monohydrate