missing translation for 'onlineSavingsMsg'
Learn More
Sodium Hydroxide, DILUT-IT Concentrate, 5N, J.T. Baker™ Product Code.: 10528912

Sodium Hydroxide, DILUT-IT Concentrate, 5N, J.T. Baker™

Product Code. 10528912
For dilution to 1 L, HDPE Sealed bottle
Click to view available options
Quantity:
For dilution to 1 L
Packaging:
HDPE Sealed bottle
This item is not returnable. View return policy

Product Code. 10528912

Brand: JT Baker 4690

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Prepare fresh, reliable standard solutions quickly and accurately. Sodium Hydroxide, DILUT-IT Concentrate, 5N, J.T. Baker™ is a pre-mixed volumetric concentrate for dilution to 1L that is ideal for titrimetry and other forms of wet chemical analysis.

DILUT-IT solutions are relied upon for fast, accurate results in life sciences and pharmaceutical quality control applications. Simply dilute the volumetric concentrate to 1 liter (carefully following instructions provided with the package) and the resulting solution will have an exact normality within ±0.1%.

  • Enhanced productivity: Eliminate expensive, time-consuming steps, such as the selection and analysis of starting reagents, and weighing and standardization of the final solution
  • Space saving: Products require only one-tenth of the space required for ready-made solutions
  • Reduced shipping costs: Spend less on shipping due to the reduced weight and package size of a concentrate

Specifications

Color Colorless
Boiling Point 100°C
CAS 1310-73-2
Melting Point 0°C
Molecular Formula HNaO
Concentration 5mol/L
Density 1.3g/cm3 at 20°C
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Fully miscible in water
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity For dilution to 1 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Grade DILUT-IT
Normality 5N
Odor Odorless
Packaging HDPE Sealed bottle
Physical Form Concentrate
Chemical Name or Material Sodium hydroxide
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hydroxide, DILUT-IT Concentrate, 5N, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.